Learn More
Cole-Parmer Cole-Parmer large Balance Draft Sheild, Large 18"W x 18-1/2"H x 19-3/4"D
Catalog No. NC0019824 Shop All Cole-Parmer Products
Encompass_Preferred
Clear acrylic prevents drafts from affecting weighings. No assembly is required. There is a mouse hole in the back panel for power cords and raised edges on top of enclosure to allow a secure placement of frequently used weighing accessories. It also minimizes air movement around your balance for faster, more stable readings.
Catalog No. NC0019824 Supplier Cole-Parmer Supplier No. 0101906
Clear acrylic prevents drafts from affecting weighings. No assembly is required. There is a mouse hole in the back panel for power cords and raised edges on top of enclosure to allow a secure placement of frequently used weighing accessories. It also minimizes air movement around your balance for faster, more stable readings.