Learn More
Cole-Parmer Essentials Balance Draft Shield, Medium 15”W x 15-1/2”H x 16-3/4”D
Catalog No. NC1858771 Shop All Cole-Parmer Products
Encompass_Preferred
Clear acrylic prevents drafts from affecting weighingsNo assembly requiredMouse hole in back panel for power cordsRaised edges on top of enclosure allow secure placement of frequently used weighing accessoriesMinimizes air movement around your balance for faster, more stable readings
Catalog No. NC1858771 Supplier Cole-Parmer Supplier No. 0101904
Clear acrylic prevents drafts from affecting weighingsNo assembly requiredMouse hole in back panel for power cordsRaised edges on top of enclosure allow secure placement of frequently used weighing accessoriesMinimizes air movement around your balance for faster, more stable readings