Learn More
Cole-Parmer Cole-Parmer small Balance Draft Shield, Small 13" W x 13-1/2" H x 14-3/4" D
Catalog No. NC9943134 Shop All Cole-Parmer Products
Encompass_Preferred
Clear acrylic prevents drafts from affecting weighings. No assembly is required. There is a mouse hole in the back panel for power cords and raised edges on top of enclosure to allow a secure placement of frequently used weighing accessories. It also minimizes air movement around your balance for faster, more stable readings.
Catalog No. NC9943134 Supplier Cole-Parmer Supplier No. 0101902
Clear acrylic prevents drafts from affecting weighings. No assembly is required. There is a mouse hole in the back panel for power cords and raised edges on top of enclosure to allow a secure placement of frequently used weighing accessories. It also minimizes air movement around your balance for faster, more stable readings.