Learn More
Yamato Scientific America, Inc. Vented Balance Enclosure 115V 48" Wide
Catalog No. NC1829078 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Self-contained laboratory unit ideal for fine powders, chemicals and biological products. Designed with top-mounted HEPA/blower filtration set up to circualte internal chamber atmosphere, through the HEPA filter and into the lab. Constructed with optically clear 3/8" jeweled acrylic plastics and phenolic base with superior chemical resistance. Outside width dimension is 48". 115V.
Catalog No. NC1829078 Supplier Yamato Scientific America, Inc. Supplier No. VBE408
Add to Cart
Add to Cart
Request a Quote
Self-contained laboratory unit ideal for fine powders, chemicals and biological products. Designed with top-mounted HEPA/blower filtration set up to circualte internal chamber atmosphere, through the HEPA filter and into the lab. Constructed with optically clear 3/8" jeweled acrylic plastics and phenolic base with superior chemical resistance. Outside width dimension is 48". 115V.