Learn More
Adam Equipment HCB 123 Highland Portable Precision Balance, 120g Capacity and 0.001g Readability
Catalog No. NC0536478
Encompass_Preferred
Adam Equipments HCB Highland Portable balances offers advanced features in a compact, durable balance that stacks for easy storage. Featuring multiple weighing applications including parts counting, percent weighing and accumulation, the Highland provides fast, accurate weighing in a number of different environments – from science research in the laboratory or field, to manufacturing operations. Sturdy ABS construction allows the Highland to stand up to demanding lab use, while Highland’s ShockProtect offers advanced overload protection to keep the balance safe. HandiCal facilitates internal calibration without the need for external weights. Highland balances are well-suited for lab work, field use, and many industrial applications. The Highland features HandiCal internal calibration with a built-in mass, and ShockProtect as a safeguard against shock loading and accidental overload.
Catalog No. NC0536478 Supplier Adam Equipment Supplier No. HCB 123
Adam Equipments HCB Highland Portable balances offers advanced features in a compact, durable balance that stacks for easy storage. Featuring multiple weighing applications including parts counting, percent weighing and accumulation, the Highland provides fast, accurate weighing in a number of different environments – from science research in the laboratory or field, to manufacturing operations. Sturdy ABS construction allows the Highland to stand up to demanding lab use, while Highland’s ShockProtect offers advanced overload protection to keep the balance safe. HandiCal facilitates internal calibration without the need for external weights. Highland balances are well-suited for lab work, field use, and many industrial applications. The Highland features HandiCal internal calibration with a built-in mass, and ShockProtect as a safeguard against shock loading and accidental overload.