Learn More
Sartorius Balance Interface Cables
Catalog No. 01913371
Change view
Click to view available options
Cable Interface D1 To Ibm At
Cable Interface D1 To Ibm Pc
Cable25pin-rj11 6ft For Xl Bal
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-913-371 Cable Interface D1 To Ibm At
01-913-353 Cable25pin-rj11 6ft For Xl Bal
01-913-370 Cable Interface D1 To Ibm Pc
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-913-371 Supplier Sartorius Supplier No. 601430.1
Interface Cables. Connect your Sartorius balance to a 9-pin connector on a computer. All cables are 1m long.