Learn More
Electron Microscopy Sciences EMS Mini Lab Balance 100g / 0.01g readability
SDP
Catalog No. 501927850
Encompass_Preferred
EMS Mini Lab Balances are extraordinarily compact, easy to use, and convenient for a variety of laboratory weighing applications. With a footprint of only 2.6 x 3.9", these Mini Balances offer exceptional accuracy and linearity through their full weighing ranges.
Available with either a 100g or 500g capacity, these digital balances feature an easy to read, backlit, liquid crystal display. Buttons for ON/OFF, MODE and TARE are located on the front for easy access. The software includes a calibration function for in-lab calibration with a 100g or 500g calibration weight. Calibration weights are available separately.
Designed for portability, EMS Mini Balances are powered by 2 AAA batteries (included). After one minute sitting idle, the power automatically shuts off to conserve battery life. Ideal for lab or field use, each balance includes a snap on, protective cover, with instructions conveniently printed on the underside.
Catalog No. 50-192-7850 Supplier Electron Microscopy Sciences Supplier No. 8705201
EMS Mini Lab Balances are extraordinarily compact, easy to use, and convenient for a variety of laboratory weighing applications. With a footprint of only 2.6 x 3.9", these Mini Balances offer exceptional accuracy and linearity through their full weighing ranges.
Available with either a 100g or 500g capacity, these digital balances feature an easy to read, backlit, liquid crystal display. Buttons for ON/OFF, MODE and TARE are located on the front for easy access. The software includes a calibration function for in-lab calibration with a 100g or 500g calibration weight. Calibration weights are available separately.
Designed for portability, EMS Mini Balances are powered by 2 AAA batteries (included). After one minute sitting idle, the power automatically shuts off to conserve battery life. Ideal for lab or field use, each balance includes a snap on, protective cover, with instructions conveniently printed on the underside.