Learn More
Hach Company Balance, Ohaus Model YA501
Supplier:  Hach Company 2796900
Encompass
Designed for portability, this scale is ideal for remote or "on the go" applications. Weighs in g, oz, ct, gn, dwt. Unique hinged cover protects the scale with auto shut-off feature. Balance measures to 0.1 g. 500 g capacity. Weigh Pan measures 2.8 in x 2 in (70 mm x 50 mm)
Catalog No. 50-199-6685