Learn More
Scientific Industries BALANCE PHARMACY CLASS II
Supplier:  Scientific Industries DRX4120
Encompass_Preferred
Class II pharmacy balance, NTEP certified as legal for trade in prescription weighing. Capacity 100g x 0.001g, with a tare capacity equal to its range, and provides a display of scale capacity used (in percent) whenever weighing.
Catalog No. 50-728-236