Learn More
Cole-Parmer Speirs Robertson AMS-30x45 AMS Balance Platform, Sorbothane Isolators; 30x45 cm
Catalog No. NC1280255 Shop All Cole-Parmer Products
Encompass
The AMS models use sorbothane isolators, which is perfect for heavy instruments. With a natural frequency of 7 Hz, the majority of building vibrations will be removed. This platform removes up to 90 percent of vibrations to improve performance of balances. Combination of isolators and sound-deadened layer construction removes both building and surface vibrations. Seamless pharma-grade (Ra finish 0.3) stainless steel worktops are chemical resistant and meet stringent cleaning requirements.
Catalog No. NC1280255 Supplier Cole-Parmer Supplier No. 1150004
The AMS models use sorbothane isolators, which is perfect for heavy instruments. With a natural frequency of 7 Hz, the majority of building vibrations will be removed. This platform removes up to 90 percent of vibrations to improve performance of balances. Combination of isolators and sound-deadened layer construction removes both building and surface vibrations. Seamless pharma-grade (Ra finish 0.3) stainless steel worktops are chemical resistant and meet stringent cleaning requirements.