Learn More
Scientific Industries BALANCE PRECISION AUTOCAL USB
Catalog No. 50728264
Encompass_Preferred
4000g x 0.01g w/ Automatic Internal Calibration, ideal for weighing small loads, Auto calibration keeps the balance calibrated at all times. The balance includes 12 weighing functions, and it is equipped with USB, RS232, and PS2 interface ports
Catalog No. 50-728-264 Supplier Scientific Industries Supplier No. AG4000
4000g x 0.01g w/ Automatic Internal Calibration, ideal for weighing small loads, Auto calibration keeps the balance calibrated at all times. The balance includes 12 weighing functions, and it is equipped with USB, RS232, and PS2 interface ports