Learn More
Thomas Scientific HighlandT Portable Precision Balance, 600g x 0.01g, 110V
SDP
Catalog No. 501458313
Encompass_Preferred
Highland balances have what it takes for basic lab work, field use and various industrial applications. Features 15 weighing units Zero Tracking Backlit LCD display Capacity tracker ShockProtect™ overload three-point protection Can be stacked for storage (120mm pan size only) Lock down mounting slot for Kensington™ type lock Removable draught shield (120mm pan size only) Stainless steel pan Non-slip adjustable levelling feet Below balance weighing with hanger Easy access to the rechargeable battery Colour coded, sealed keypad Dual tare keys RS-232 and USB interface Handical™ manual internal calibration with built-in mass External calibration Auto power off Full range tare Low battery indication AC Adapter Platform Diameter: 4.7" (120 mm) Applications Weighing Percentage weighing Parts counting Below balance weighing Weight accumulation
Catalog No. 50-145-8313 Supplier Thomas Scientific Supplier No. 1224C66
Highland balances have what it takes for basic lab work, field use and various industrial applications. Features 15 weighing units Zero Tracking Backlit LCD display Capacity tracker ShockProtect™ overload three-point protection Can be stacked for storage (120mm pan size only) Lock down mounting slot for Kensington™ type lock Removable draught shield (120mm pan size only) Stainless steel pan Non-slip adjustable levelling feet Below balance weighing with hanger Easy access to the rechargeable battery Colour coded, sealed keypad Dual tare keys RS-232 and USB interface Handical™ manual internal calibration with built-in mass External calibration Auto power off Full range tare Low battery indication AC Adapter Platform Diameter: 4.7" (120 mm) Applications Weighing Percentage weighing Parts counting Below balance weighing Weight accumulation