Learn More
Uline OHAUS Scout Balance Scale 2,200g x 0.01g
Catalog No. NC2165796
Encompass
Balance scale, 2,200g x 0.01g, platform size: 6.7 x 5.5"""", powered by 4 AA batteries or included AC adapter
This item is not returnable. View return policy
Catalog No. NC2165796 Supplier Uline Supplier No. H7294
Balance scale, 2,200g x 0.01g, platform size: 6.7 x 5.5"""", powered by 4 AA batteries or included AC adapter