Learn More
Grainger A&D Weighing Precision Balance, 120 g Capacity, 0.01 g Resolution
Catalog No. 502779138
Encompass_Preferred
The EK-120i is a compact, high-precision bench scale designed for versatile weighing in laboratory and industrial settings. Built with a rugged metal housing and a stainless steel weighing pan with 110 mm dia provides reliable performance through integrated SG weighing technology. The balance features a high-visibility, backlit 16 mm LCD for clear readings and supports multiple applications including counting, percentage, and comparator tasks.
  • Capacity of 120 g with 0.01 g readability
  • Comparator mode with HI, OK, and LO annunciation
  • Counting function with automatic counting accuracy improvement
  • Conforms to GLP, GMP, and ISO standards
  • RS-232C interface for data communication
  • External calibration with user-selectable mass
  • Operating temperature range of -10°C to 40°C
  • Flexible power via AC adapter or optional Ni-MH battery
Catalog No. 50-277-9138 Supplier Grainger Supplier No. 19NC65
The EK-120i is a compact, high-precision bench scale designed for versatile weighing in laboratory and industrial settings. Built with a rugged metal housing and a stainless steel weighing pan with 110 mm dia provides reliable performance through integrated SG weighing technology. The balance features a high-visibility, backlit 16 mm LCD for clear readings and supports multiple applications including counting, percentage, and comparator tasks.
  • Capacity of 120 g with 0.01 g readability
  • Comparator mode with HI, OK, and LO annunciation
  • Counting function with automatic counting accuracy improvement
  • Conforms to GLP, GMP, and ISO standards
  • RS-232C interface for data communication
  • External calibration with user-selectable mass
  • Operating temperature range of -10°C to 40°C
  • Flexible power via AC adapter or optional Ni-MH battery