Learn More
Grainger A&D Weighing Washdown Platform Scale, Stainless Steel, 13 lb Capacity, Small Platform, 1 ea
Catalog No. 502779861
Encompass_Preferred
The SW-6KSF is an advanced washdown platform scale designed for food processing and demanding environments where hygiene is critical. Withstands high-pressure-1,450 psi and high-temperature (175°F) cleaning to ensure sanitation. The unit is constructed with a 304 stainless steel platform and frame, paired with a hermetically sealed 630 stainless steel load cell for reliability in damp locations.
  • IP69K-rated for superior dust and water resistance
  • Constructed with SUS 304 stainless steel platform and frame
  • Bright, multicolored LED display with a five-position comparator function for checkweighing
  • Integrated counting function to streamline weighing processes
  • Touch-sensitive, polycarbonate keys for durable operation
  • NTEP certified and NSF listed to meet HACCP requirements
  • Adjustable indicator bracket for desktop or wall-mount options
Catalog No. 50-277-9861 Supplier Grainger Supplier No. 19ND08
The SW-6KSF is an advanced washdown platform scale designed for food processing and demanding environments where hygiene is critical. Withstands high-pressure-1,450 psi and high-temperature (175°F) cleaning to ensure sanitation. The unit is constructed with a 304 stainless steel platform and frame, paired with a hermetically sealed 630 stainless steel load cell for reliability in damp locations.
  • IP69K-rated for superior dust and water resistance
  • Constructed with SUS 304 stainless steel platform and frame
  • Bright, multicolored LED display with a five-position comparator function for checkweighing
  • Integrated counting function to streamline weighing processes
  • Touch-sensitive, polycarbonate keys for durable operation
  • NTEP certified and NSF listed to meet HACCP requirements
  • Adjustable indicator bracket for desktop or wall-mount options