Learn More
Grainger A&D Weighing Washdown Platform Scale, Stainless Steel, 130lb Capacity
Catalog No. 502779177
Encompass_Preferred
The SW-60KMF is a high-pressure washdown bench scale engineered for demanding environments where hygiene and sanitation are essential. With a robust IP69K-rated stainless steel frame, load cell, and weigh pan, this digital scale is ideal for food processing and damp locations. It delivers fast, reliable weighing with a 1-second stabilization time and features a bright LED display with a five-position comparator function for efficient checkweighing.
  • IP69K and IP68 compliant construction supports high-pressure and high-temperature cleaning
  • Stainless steel 304 weigh pan and frame with hermetically sealed load cell
  • Legal for Trade certification (NTEP and Measurement Canada)
  • Flexible weighing units including kg, g, lb, and oz
  • Capacitive touch keys with adjustable sensitivity
  • Supports AC power or optional battery with 90 hr. of operation
Catalog No. 50-277-9177 Supplier Grainger Supplier No. 19ND12
The SW-60KMF is a high-pressure washdown bench scale engineered for demanding environments where hygiene and sanitation are essential. With a robust IP69K-rated stainless steel frame, load cell, and weigh pan, this digital scale is ideal for food processing and damp locations. It delivers fast, reliable weighing with a 1-second stabilization time and features a bright LED display with a five-position comparator function for efficient checkweighing.
  • IP69K and IP68 compliant construction supports high-pressure and high-temperature cleaning
  • Stainless steel 304 weigh pan and frame with hermetically sealed load cell
  • Legal for Trade certification (NTEP and Measurement Canada)
  • Flexible weighing units including kg, g, lb, and oz
  • Capacitive touch keys with adjustable sensitivity
  • Supports AC power or optional battery with 90 hr. of operation