Learn More
Grainger A&D Weighing Platform Bench Scale, Stainless Steel, 130 lb, 0.02 lb Readability, 1 ea
Catalog No. 502783355
Encompass_Preferred
The SE-60KAL is a digital platform bench scale engineered for demanding industrial environments. Featuring a stainless steel weighing pan and a waterproof, corrosion-resistant load cell, this unit maintains high precision with a 130 lb (60 kg) capacity and 0.02 lb (0.01 kg) readability. It holds an IP68 rating for protection against dust and water, and is approved for Legal for Trade use.
  • Supersized 7-segment LCD with 39 mm high characters
  • Stabilization speed of 1 second
  • Battery operation using 6 D-cells for up to 500 hours
  • Multiple weighing units including kg, lb, oz, and pieces
  • External calibration capability with user-selectable weight
  • Optional quick USB or RS-232C interfaces with comparator relay output
  • Operating temperature range of -10°C to 40°C (14°F to 104°F)
Catalog No. 50-278-3355 Supplier Grainger Supplier No. 19ND29
The SE-60KAL is a digital platform bench scale engineered for demanding industrial environments. Featuring a stainless steel weighing pan and a waterproof, corrosion-resistant load cell, this unit maintains high precision with a 130 lb (60 kg) capacity and 0.02 lb (0.01 kg) readability. It holds an IP68 rating for protection against dust and water, and is approved for Legal for Trade use.
  • Supersized 7-segment LCD with 39 mm high characters
  • Stabilization speed of 1 second
  • Battery operation using 6 D-cells for up to 500 hours
  • Multiple weighing units including kg, lb, oz, and pieces
  • External calibration capability with user-selectable weight
  • Optional quick USB or RS-232C interfaces with comparator relay output
  • Operating temperature range of -10°C to 40°C (14°F to 104°F)