Learn More
Grainger A&D Weighing FG-150KBM Digital Bench Scale, 300 lb, Stainless Steel Platform, 1 ea
Catalog No. 502779866
Encompass_Preferred
This compact bench scale is designed for basic counting and checkweighing applications in dry environments. Featuring a digital interface and a stainless steel weighing platform, it provides precise measurements with a 300 lb capacity.
  • Capacity of 300 lb (150 kg) with 0.02 lb (0.01 kg) readability.
  • Equipped with an RS-232C port for data connectivity.
  • Features a 7-segment LCD display that updates 10 times per second.
  • Includes checkweighing functionality for over/under limit indication.
  • Operates on an AC adapter or 4 C-size batteries for up to 150 hours of use.
  • Supports weighing in lb, kg, and oz units.
  • Stable one-second response time.
Catalog No. 50-277-9866 Supplier Grainger Supplier No. 19ND02
This compact bench scale is designed for basic counting and checkweighing applications in dry environments. Featuring a digital interface and a stainless steel weighing platform, it provides precise measurements with a 300 lb capacity.
  • Capacity of 300 lb (150 kg) with 0.02 lb (0.01 kg) readability.
  • Equipped with an RS-232C port for data connectivity.
  • Features a 7-segment LCD display that updates 10 times per second.
  • Includes checkweighing functionality for over/under limit indication.
  • Operates on an AC adapter or 4 C-size batteries for up to 150 hours of use.
  • Supports weighing in lb, kg, and oz units.
  • Stable one-second response time.