Learn More
Grainger A&D Weighing Compact Bench Scale, 310 g Capacity, 0.1 g Readability, Stainless Steel Pan, 1 ea
Catalog No. 502779870
Encompass_Preferred
The HT Series portable bench scale is a versatile, digital weighing instrument designed for applications in laboratories, schools, and general precision weighing. It provides reliable performance with a 310 g capacity and 0.1 g readability, featuring a durable stainless steel weighing pan and a clear, backlit LCD display for ease of use in various lighting conditions.
  • Capacity: 310 g with 0.1 g resolution
  • Stainless steel weighing pan
  • Backlit LCD display with 14 mm character height
  • Multiple weighing units including g, N, oz, ozt, and tl
  • Operates on 4 AA batteries (included) or optional AC adapter
  • Stackable storage case with support for weighing while in the case
  • Integrated Kensington security lock slot
  • Automatic power-off function with 5-minute inactivity timer
  • Stable, net, and zero weight indicators
Catalog No. 50-277-9870 Supplier Grainger Supplier No. 19ND50
The HT Series portable bench scale is a versatile, digital weighing instrument designed for applications in laboratories, schools, and general precision weighing. It provides reliable performance with a 310 g capacity and 0.1 g readability, featuring a durable stainless steel weighing pan and a clear, backlit LCD display for ease of use in various lighting conditions.
  • Capacity: 310 g with 0.1 g resolution
  • Stainless steel weighing pan
  • Backlit LCD display with 14 mm character height
  • Multiple weighing units including g, N, oz, ozt, and tl
  • Operates on 4 AA batteries (included) or optional AC adapter
  • Stackable storage case with support for weighing while in the case
  • Integrated Kensington security lock slot
  • Automatic power-off function with 5-minute inactivity timer
  • Stable, net, and zero weight indicators