Learn More
Grainger A&D Weighing Food Scale, IP69K, NTEP, Stainless Steel, 1 ea
Catalog No. 502779783
Encompass_Preferred
The SW series washdown scales are designed for high-sanitation, high-pressure environments, including food processing facilities. These digital scales feature a hermetically sealed load cell and stainless steel construction, ensuring durability and simplified cleaning in wet or dusty settings. The system offers versatile functionality for precision weighing tasks, supporting operations in demanding industrial conditions.
  • IP69K-rated protection against high-pressure water and extreme temperatures
  • 304 stainless steel weighing pan and frame for corrosion resistance
  • Bright, multicolored LED display with a five-position comparator
  • Integrated functions for parts counting and checkweighing
  • Legal for trade certification from NTEP and Measurement Canada
  • Fast stabilization time of approximately 1 sec.
  • RS-232C connectivity for data management
Catalog No. 50-277-9783 Supplier Grainger Supplier No. 19ND10
The SW series washdown scales are designed for high-sanitation, high-pressure environments, including food processing facilities. These digital scales feature a hermetically sealed load cell and stainless steel construction, ensuring durability and simplified cleaning in wet or dusty settings. The system offers versatile functionality for precision weighing tasks, supporting operations in demanding industrial conditions.
  • IP69K-rated protection against high-pressure water and extreme temperatures
  • 304 stainless steel weighing pan and frame for corrosion resistance
  • Bright, multicolored LED display with a five-position comparator
  • Integrated functions for parts counting and checkweighing
  • Legal for trade certification from NTEP and Measurement Canada
  • Fast stabilization time of approximately 1 sec.
  • RS-232C connectivity for data management