Learn More
Grainger A&D Weighing Shipping Scale, 400 lb Capacity, Stainless Steel Platform, Column-Mounted, 1 ea
Catalog No. 502783339
Encompass_Preferred
This versatile, high-resolution scale is designed for efficient bench-top or floor-based weighing applications. It features a robust painted steel frame with a 430 stainless steel pan, offering durability in dry environments. The unit provides multi-functional performance, including checkweighing with Hi/Ok/Lo comparator indication and parts counting capabilities.
  • Capacity of 400 lb (200 kg) with selectable weighing units in lb, kg, and oz.
  • Equipped with an easy-to-read 7-segment LCD display featuring 26 mm character height.
  • Supports AC adapter or 4 C-cell battery operation, with approximately 150 hours of battery life.
  • Includes an auto-power off function and low power alert to optimize battery usage.
  • Offers flexible operation with an integrated level bubble and adjustable leveling feet for precision.
  • Features four intuitive keys for simple control: on/off, zero, print, and mode.
Catalog No. 50-278-3339 Supplier Grainger Supplier No. 19ND03
This versatile, high-resolution scale is designed for efficient bench-top or floor-based weighing applications. It features a robust painted steel frame with a 430 stainless steel pan, offering durability in dry environments. The unit provides multi-functional performance, including checkweighing with Hi/Ok/Lo comparator indication and parts counting capabilities.
  • Capacity of 400 lb (200 kg) with selectable weighing units in lb, kg, and oz.
  • Equipped with an easy-to-read 7-segment LCD display featuring 26 mm character height.
  • Supports AC adapter or 4 C-cell battery operation, with approximately 150 hours of battery life.
  • Includes an auto-power off function and low power alert to optimize battery usage.
  • Offers flexible operation with an integrated level bubble and adjustable leveling feet for precision.
  • Features four intuitive keys for simple control: on/off, zero, print, and mode.