The SE-30KAM is a high-performance washdown bench scale designed for reliable use in wet environments. Featuring an IP68-rated construction, this scale includes a durable stainless steel weighing surface and a waterproof, corrosion-resistant load cell, making it ideal for demanding industrial and food processing applications. Its intuitive design includes a supersized, 39 mm character height LCD for easy viewing and supports multiple weighing units, including kg, lb, oz, and pcs.
- IP68-rated dust and water protection
- Fast one-second stabilization time
- Extended battery life providing up to 4,500 hours of continuous operation on 6 D-cell batteries
- Integrated comparator function for high, OK, and low weight checking
- NTEP and Measurement Canada approved for legal-for-trade applications
- Versatile connectivity options including RS-232 and optional USB