Learn More
Mitchell Plastics Balance Shield Guards
Catalog No. 05723477
Change view
Click to view available options
For Use With (Equipment):
Mettler Balance
Sartorius Balances
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
05-723-477 Sartorius Balances
05-723-476 Mettler Balance
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-723-477 Supplier Mitchell Plastics Supplier No. RP790
  • Protect samples during weighing
  • Enclose balance leaving controls exposed for easy access
  • Hinged lid offers direct access to weighing pan
  • 1 in. hole on top allows for sample dispensing
  • Designed for Mettler and Sartorius balances
Specifications
For Use With (Equipment) Sartorius Balances
Height (English) 6.5 in.
Height (Metric) 16.51 cm