Learn More
Foxx Life Sciences EZBio Stainless Steel Balance Spoon Polished with Steel Handle, 304 SS, 175mm, 1/EA
Catalog No. NC1795101
Encompass_Preferred
Foxx Life Sciences Stainless Steel Balance Spoons are designed for ease of use and flowability. These ideal products make it possible to support numerous processes, such as formulation work, measuring, sample gathering, powder prep, granulate work, with a variety of media including pastes and liquids. Manufactured with a single piece of high-quality 304 Stainless Steel, they are free of edges, crevices, or undercuts, to eliminate the possibility of cross contamination, product residue, or cleaning agent deposits. They are perfect for uses in laboratories and in several industries including work with chemistry, pharmaceuticals, cosmetics, and food. 304 Stainless Steel High-gloss polishedEasy to clean hygienic design
Catalog No. NC1795101 Supplier Foxx Life Sciences Supplier No. 25C3601FLS
Foxx Life Sciences Stainless Steel Balance Spoons are designed for ease of use and flowability. These ideal products make it possible to support numerous processes, such as formulation work, measuring, sample gathering, powder prep, granulate work, with a variety of media including pastes and liquids. Manufactured with a single piece of high-quality 304 Stainless Steel, they are free of edges, crevices, or undercuts, to eliminate the possibility of cross contamination, product residue, or cleaning agent deposits. They are perfect for uses in laboratories and in several industries including work with chemistry, pharmaceuticals, cosmetics, and food. 304 Stainless Steel High-gloss polishedEasy to clean hygienic design