Learn More
Electron Microscopy Sciences Balance Spoon
SDP
Catalog No. 5024621
Encompass_Preferred
Stainless steel with "PTFE" coating. One end has a square trough, which holds about 1cc. 26.
Catalog No. 50-246-21 Supplier Electron Microscopy Sciences Supplier No. 72983
Stainless steel with "PTFE" coating. One end has a square trough, which holds about 1cc. 26.