Learn More
Cole-Parmer Speirs Robertson AMD-SB-90x75 AMD-SB Balance Table, Sorbothane Isolators; 90x75 cm
Catalog No. NC1673342 Shop All Cole-Parmer Products
Encompass
Creates the best environment for your balance and by optimising user comfort improves both accuracy and efficiency of measurements. The range of balance tables have been carefully designed to remove vibrations which limit balance performance and provide a tailored working environment for the user. The custom designed Sorobthane isolators are extremely efficient in removing vibrations. The stainless worktops meet stringent lab cleanliness requirements. Options include acoustic hoods, armrest bars, shelves and monitor stands, allowing you to configure a table to suit you specific needs. Table are simple to install and require no maintenance.
Catalog No. NC1673342 Supplier Cole-Parmer Supplier No. 1150027
Creates the best environment for your balance and by optimising user comfort improves both accuracy and efficiency of measurements. The range of balance tables have been carefully designed to remove vibrations which limit balance performance and provide a tailored working environment for the user. The custom designed Sorobthane isolators are extremely efficient in removing vibrations. The stainless worktops meet stringent lab cleanliness requirements. Options include acoustic hoods, armrest bars, shelves and monitor stands, allowing you to configure a table to suit you specific needs. Table are simple to install and require no maintenance.