Learn More
FUJIFILM Biosciences BalanCD CHO Growth A Medium - Liquid
Catalog No. NC1541022
Encompass
BalanCD CHO Growth A is a chemically-defined, animal component-free growth medium designed to increase process yields of antibodies and recombinant proteins in Chinese Hamster Ovary (CHO) cells. This formulation was developed using FUJIFILM Irvine Scientific’s Rational Culture Media Design strategy to achieve enhanced performance of growth and production in batch cultures.
Store at 2–8°C
Catalog No. NC1541022 Supplier FUJIFILM Biosciences Supplier No. 911281L
BalanCD CHO Growth A is a chemically-defined, animal component-free growth medium designed to increase process yields of antibodies and recombinant proteins in Chinese Hamster Ovary (CHO) cells. This formulation was developed using FUJIFILM Irvine Scientific’s Rational Culture Media Design strategy to achieve enhanced performance of growth and production in batch cultures.
Store at 2–8°C