Learn More
Supertek Scientific Balancing Bird
GREENER_CHOICESCI_ED
Catalog No. S81671 Shop All Supertek Scientific Products
Change view
Click to view available options
Dimensions (L x W x H):
6 x 6 x 2.5 in.
Catalog No. Dimensions (L x W x H)
S81671 6 x 6 x 2.5 in.
1 options
Catalog No. S81671 Supplier Supertek Scientific Supplier No. PH10560SP
Add to Cart
Add to Cart
Demonstrates how gravity works
  • This simple device demonstrates how gravity works
  • By perfectly distributing weight on the tips of the wings, and placing its mass near the tip of its beak, the equilibrium of the bird is stabilized
  • This allows the bird to balance on whatever object it is place upon
Specifications
Product Type Balancing Bird
For Use With (Application) Demonstrate Gravity
Dimensions (L x W x H) 6 x 6 x 2.5 in.