Learn More
MSC Balax Plug Thread Insert Go/No Go Gage, 6-32 Thread, Class 3B, High-Speed Tool Steel
Catalog No. 5000532942
Encompass
This Go/No Go plug thread insert gage is designed for accurately checking 6-32 thread sizes with a 3B classification. Made from high-speed tool steel, it ensures precision and durability for quality control applications involving screw thread inserts (STI).
  • Accurately checks 6-32 thread sizes.
  • Conforms to 3B classification standards.
  • Constructed from high-speed tool steel for durability.
  • Designed for use with screw thread inserts.
  • Provides both 'Go' and 'No Go' measurements for comprehensive thread inspection.
Catalog No. 50-005-32942 Supplier MSC Supplier No. 05349121
This Go/No Go plug thread insert gage is designed for accurately checking 6-32 thread sizes with a 3B classification. Made from high-speed tool steel, it ensures precision and durability for quality control applications involving screw thread inserts (STI).
  • Accurately checks 6-32 thread sizes.
  • Conforms to 3B classification standards.
  • Constructed from high-speed tool steel for durability.
  • Designed for use with screw thread inserts.
  • Provides both 'Go' and 'No Go' measurements for comprehensive thread inspection.