Learn More
MSC Balax Screw Thread Insert Go/No Go Gage, 10-32 Thread, High-Speed Tool Steel, Class 2B
Catalog No. 5000516863
Encompass
This go/no go gage is specifically designed for inspecting screw threads with a 10-32 thread size. Constructed from high-speed tool steel, it ensures durability and precision for critical measurements. It features a 2B classification and is double-ended with an integrated handle, making it suitable for various industrial applications.
  • Checks 10-32 thread size.
  • Constructed from high-speed tool steel.
  • Features a 2B classification.
  • Designed as a screw thread insert (STI) gage.
  • Equipped with go and no go ends.
  • Includes a handle for easy use.
  • Conforms to ANSI standards.
Catalog No. 50-005-16863 Supplier MSC Supplier No. 05349238
This go/no go gage is specifically designed for inspecting screw threads with a 10-32 thread size. Constructed from high-speed tool steel, it ensures durability and precision for critical measurements. It features a 2B classification and is double-ended with an integrated handle, making it suitable for various industrial applications.
  • Checks 10-32 thread size.
  • Constructed from high-speed tool steel.
  • Features a 2B classification.
  • Designed as a screw thread insert (STI) gage.
  • Equipped with go and no go ends.
  • Includes a handle for easy use.
  • Conforms to ANSI standards.