Learn More
iQ Biosciences BALB/c Mouse PBMCs, 5M cells/vial
Catalog No. NC0999599
Encompass_Preferred
High-quality, high-viability (> 80%) peripheral blood mononuclear cells (PBMCs) from the BALB/c mouse, a commonly-used inbred mouse strain. Used for a wide variety of immunology-based applications, including in-vitro immunoassays and ex-vivo applications. All orders come with an iQ Certificate of Analysis.
Catalog No. NC0999599 Supplier iQ Biosciences Supplier No. IQBMPB101
High-quality, high-viability (> 80%) peripheral blood mononuclear cells (PBMCs) from the BALB/c mouse, a commonly-used inbred mouse strain. Used for a wide variety of immunology-based applications, including in-vitro immunoassays and ex-vivo applications. All orders come with an iQ Certificate of Analysis.