Learn More
MSC Baldwin Cab Air Element, 16 3/8 x 8 9/16 x 1 9/16 in
Catalog No. 5000509889
Encompass
Panel Cabin Air Filters are designed to prevent contaminants and pollutants from entering the cabin area. Their space-saving design makes them suitable for a wide variety of heavy-duty trucks and passenger vehicles.
  • Panel cabin air filters
  • Space-saving design
  • Attached gasket for secure fit
  • Helps prevent contaminants and pollutants from entering the cabin area
  • Utilized in a wide variety of heavy-duty trucks and passenger vehicles
  • Suitable for John Deere Loaders and Tool Carriers
Catalog No. 50-005-09889 Supplier MSC Supplier No. 55873871
Panel Cabin Air Filters are designed to prevent contaminants and pollutants from entering the cabin area. Their space-saving design makes them suitable for a wide variety of heavy-duty trucks and passenger vehicles.
  • Panel cabin air filters
  • Space-saving design
  • Attached gasket for secure fit
  • Helps prevent contaminants and pollutants from entering the cabin area
  • Utilized in a wide variety of heavy-duty trucks and passenger vehicles
  • Suitable for John Deere Loaders and Tool Carriers