Learn More
MSC Baldwin Channel Flow Air Element, 12 1/16 in, 1 ea
Catalog No. 5000506448
Encompass
Channel Flow air filter elements ensure proper equipment function by preventing airborne contaminants from reaching the combustion chamber.
  • Channel Flow air element
  • Air filter style
  • Outside diameter 12 1/16 in (306.4 mm)
  • Length 8 3/32 in (205.6 mm)
  • Attached gasket included
  • Three bolts included
Catalog No. 50-005-06448 Supplier MSC Supplier No. 19375559
Channel Flow air filter elements ensure proper equipment function by preventing airborne contaminants from reaching the combustion chamber.
  • Channel Flow air element
  • Air filter style
  • Outside diameter 12 1/16 in (306.4 mm)
  • Length 8 3/32 in (205.6 mm)
  • Attached gasket included
  • Three bolts included