Learn More
MSC Baldwin Channel Flow Air Element, 9 5/16 in, 1 ea
Catalog No. 5000506439
Encompass
Baldwin Channel Flow air filter elements prevent airborne contaminants from reaching the combustion chamber, ensuring proper equipment function.
  • Designed as a channel flow air element
  • Features an attached gasket
  • Suitable for Ford Excursion and Super Duty Trucks with 6.0L Turbo Diesel Engine applications
  • Has a length of 9 5/16 in (236.5 mm)
  • Features an outside diameter range of 7 31/32 in (202.4 mm) to 12 3/32 in (307.2 mm)
Catalog No. 50-005-06439 Supplier MSC Supplier No. 19375419
Baldwin Channel Flow air filter elements prevent airborne contaminants from reaching the combustion chamber, ensuring proper equipment function.
  • Designed as a channel flow air element
  • Features an attached gasket
  • Suitable for Ford Excursion and Super Duty Trucks with 6.0L Turbo Diesel Engine applications
  • Has a length of 9 5/16 in (236.5 mm)
  • Features an outside diameter range of 7 31/32 in (202.4 mm) to 12 3/32 in (307.2 mm)