Baldwin Panel Air Filters are designed to protect vehicles and equipment by efficiently removing airborne contaminants from the intake air of combustion engines. These filters enhance engine performance and longevity, constructed with premium materials to ensure optimal airflow while trapping pollutants. They are suitable for a range of vehicles and machinery, engineered for compatibility with specific engine types, and provide reliable operation even in demanding environments.
- Set of two DynaCell panel air elements
- Each filter measures 45 3/8 in x 14 in x 6 3/16 in (1152.5 mm x 355.6 mm x 157.2 mm)
- Uses two filters per application
- Utilizes filtration technology
- Suitable for industrial applications
- Ensures optimal airflow
- Provides reliable operation in demanding environments