Learn More
MSC Baldwin Fuel Element, 2 7/8 in OD, 9.8 Nominal Micron Rating
Catalog No. 5000509659
Encompass
Modern fuel injection systems require fuel to be free of both particulate and water contamination. This fuel element is designed to keep fuel clean and engines running at maximum efficiency.
  • Designed to keep fuel clean in modern fuel injection systems.
  • Ensures engines run at maximum efficiency.
  • Outside diameter of 2 7/8 in (73.0 mm).
  • Inside diameter of 15/16 in (23.8 mm).
  • Length of 3 1/16 in (77.8 mm).
  • Includes a furnished gasket.
  • Features a micron rating of 9.8 nominal and 27 absolute.
  • Applicable in compressor and various heavy equipment systems.
Catalog No. 50-005-09659 Supplier MSC Supplier No. 55868855
Modern fuel injection systems require fuel to be free of both particulate and water contamination. This fuel element is designed to keep fuel clean and engines running at maximum efficiency.
  • Designed to keep fuel clean in modern fuel injection systems.
  • Ensures engines run at maximum efficiency.
  • Outside diameter of 2 7/8 in (73.0 mm).
  • Inside diameter of 15/16 in (23.8 mm).
  • Length of 3 1/16 in (77.8 mm).
  • Includes a furnished gasket.
  • Features a micron rating of 9.8 nominal and 27 absolute.
  • Applicable in compressor and various heavy equipment systems.