Learn More
MSC Baldwin Fuel Element, Diesel, 25 Absolute Micron, Attached Gasket
Catalog No. 5000508418
Encompass
This fuel element is designed to ensure modern fuel injection systems are free of particulate and water contamination. It helps keep fuel clean and engines running at maximum efficiency.
  • Helps keep fuel clean for optimal engine performance.
  • Ensures fuel injection systems are free of particulate and water contamination.
  • Provides 5 nominal and 25 absolute micron filtration.
  • Features an attached gasket.
  • Designed for use in Hitachi and Komatsu equipment.
  • Outside diameter of 2 7/8 in (73.0 mm).
  • Inside diameter of 9/16 in (14.3 mm).
  • Length of 4 11/32 in (110.3 mm).
Catalog No. 50-005-08418 Supplier MSC Supplier No. 55866164
This fuel element is designed to ensure modern fuel injection systems are free of particulate and water contamination. It helps keep fuel clean and engines running at maximum efficiency.
  • Helps keep fuel clean for optimal engine performance.
  • Ensures fuel injection systems are free of particulate and water contamination.
  • Provides 5 nominal and 25 absolute micron filtration.
  • Features an attached gasket.
  • Designed for use in Hitachi and Komatsu equipment.
  • Outside diameter of 2 7/8 in (73.0 mm).
  • Inside diameter of 9/16 in (14.3 mm).
  • Length of 4 11/32 in (110.3 mm).