Learn More
MSC Baldwin Fuel Spin-On, 10 Micron, M72 x 2.0, 1 ea
Catalog No. 5000505370
Encompass
Modern fuel injection systems require fuel be free of both particulate and water contamination. This spin-on fuel filter keeps fuel clean and engines running at maximum efficiency.
  • Spin-on fuel filter type
  • Integral gasket included
  • Micron rating: 10 absolute
  • Outside diameter 3 23/32 in (94.5 mm)
  • Length 9 5/32 in (232.6 mm)
  • Thread size: M72 x 2.0
Catalog No. 50-005-05370 Supplier MSC Supplier No. 10954568
Modern fuel injection systems require fuel be free of both particulate and water contamination. This spin-on fuel filter keeps fuel clean and engines running at maximum efficiency.
  • Spin-on fuel filter type
  • Integral gasket included
  • Micron rating: 10 absolute
  • Outside diameter 3 23/32 in (94.5 mm)
  • Length 9 5/32 in (232.6 mm)
  • Thread size: M72 x 2.0