Learn More
MSC Baldwin Fuel Spin-on, 5/8-11 Thread, 4 Micron, International Engines
Catalog No. 5000509798
Encompass
These fuel spin-on filters are designed to ensure fuel remains free of particulate and water contamination, which is crucial for modern fuel injection systems. They are engineered to keep engines running at maximum efficiency.
  • Keeps fuel clean
  • Ensures engines run at maximum efficiency
  • Removes particulate and water contamination
  • Designed for modern fuel injection systems
Catalog No. 50-005-09798 Supplier MSC Supplier No. 55863807
These fuel spin-on filters are designed to ensure fuel remains free of particulate and water contamination, which is crucial for modern fuel injection systems. They are engineered to keep engines running at maximum efficiency.
  • Keeps fuel clean
  • Ensures engines run at maximum efficiency
  • Removes particulate and water contamination
  • Designed for modern fuel injection systems