Learn More
MSC Baldwin Fuel Spin-on, M20 x 1.5 Thread, 4 Nominal Micron, Integral Post Seal
Catalog No. 5000508373
Encompass
Modern fuel injection systems require fuel to be free of both particulate and water contamination. These spin-on fuel filters keep fuel clean and engines running at maximum efficiency.
  • Contains integral post seal
  • Features M20 x 1.5 thread size
  • Outside diameter of 3 11/16 in (93.7 mm)
  • Length of 5 5/8 in (142.9 mm)
  • Integral gasket
  • Micron rating of 4 nominal; 13 absolute
  • Suitable for Volvo Excavators applications
Catalog No. 50-005-08373 Supplier MSC Supplier No. 55863716
Modern fuel injection systems require fuel to be free of both particulate and water contamination. These spin-on fuel filters keep fuel clean and engines running at maximum efficiency.
  • Contains integral post seal
  • Features M20 x 1.5 thread size
  • Outside diameter of 3 11/16 in (93.7 mm)
  • Length of 5 5/8 in (142.9 mm)
  • Integral gasket
  • Micron rating of 4 nominal; 13 absolute
  • Suitable for Volvo Excavators applications