Learn More
MSC Baldwin Fuel Spin-on, M24 x 1.5 Thread, 5 Micron, Heavy Duty
Catalog No. 5000509619
Encompass
Modern fuel injection systems require fuel to be free of both particulate and water contamination. These spin-on fuel filters are designed to keep fuel clean and engines running at maximum efficiency.
  • M24 x 1.5 thread size
  • 3 11/16 in (93.7 mm) outside diameter
  • 5 21/32 in (143.7 mm) length
  • Integral gasket included
  • 5 micron absolute rating
  • Suitable for generator sets with Scania DC12 engines and trucks
Catalog No. 50-005-09619 Supplier MSC Supplier No. 55863724
Modern fuel injection systems require fuel to be free of both particulate and water contamination. These spin-on fuel filters are designed to keep fuel clean and engines running at maximum efficiency.
  • M24 x 1.5 thread size
  • 3 11/16 in (93.7 mm) outside diameter
  • 5 21/32 in (143.7 mm) length
  • Integral gasket included
  • 5 micron absolute rating
  • Suitable for generator sets with Scania DC12 engines and trucks