Learn More
MSC Baldwin Fuel/Water Separator, 1/2-20, 5 Micron, Drain, Sensor Port
Catalog No. 5000508226
Encompass
This fuel/water separator spin-on filter is designed for effective fuel filtration in heavy-duty applications, particularly for Caterpillar Engines. It features a 5 absolute micron rating to ensure clean fuel delivery and protect engine components from contaminants. The filter is equipped with both a drain and a sensor port for convenient maintenance and monitoring.
  • Spin-on fuel/water separator design
  • 5 absolute micron rating for efficient filtration
  • Equipped with a drain for water removal
  • Includes a sensor port for monitoring fuel quality
  • Thread size: 1/2-20
  • Outside diameter: 3 11/16 in (93.7 mm)
  • Length: 9 17/32 in (242.1 mm)
  • Furnished gasket (G401) and O-ring included
  • Suitable for Caterpillar Engines
Catalog No. 50-005-08226 Supplier MSC Supplier No. 55863211
This fuel/water separator spin-on filter is designed for effective fuel filtration in heavy-duty applications, particularly for Caterpillar Engines. It features a 5 absolute micron rating to ensure clean fuel delivery and protect engine components from contaminants. The filter is equipped with both a drain and a sensor port for convenient maintenance and monitoring.
  • Spin-on fuel/water separator design
  • 5 absolute micron rating for efficient filtration
  • Equipped with a drain for water removal
  • Includes a sensor port for monitoring fuel quality
  • Thread size: 1/2-20
  • Outside diameter: 3 11/16 in (93.7 mm)
  • Length: 9 17/32 in (242.1 mm)
  • Furnished gasket (G401) and O-ring included
  • Suitable for Caterpillar Engines