Learn More
MSC Baldwin Fuel/Water Separator, 5 Micron, 1/2-20 Sensor Port, Spin-on
Catalog No. 5000508216
Encompass
This fuel/water separator is designed for modern fuel injection systems, ensuring fuel is free of particulate and water contamination. It helps keep engines running at maximum efficiency.
  • Spin-on design with sensor port and drain
  • Integral post seal
  • Sensor port thread: 1/2-20
  • Filter thread size: 1-12
  • Outside diameter: 108.7 mm (4.28 in)
  • Length: 285.0 mm (11.22 in)
  • Integral gasket
  • Included o-ring
  • 5 micron absolute rating
  • Designed for John Deere equipment
Catalog No. 50-005-08216 Supplier MSC Supplier No. 55862387
This fuel/water separator is designed for modern fuel injection systems, ensuring fuel is free of particulate and water contamination. It helps keep engines running at maximum efficiency.
  • Spin-on design with sensor port and drain
  • Integral post seal
  • Sensor port thread: 1/2-20
  • Filter thread size: 1-12
  • Outside diameter: 108.7 mm (4.28 in)
  • Length: 285.0 mm (11.22 in)
  • Integral gasket
  • Included o-ring
  • 5 micron absolute rating
  • Designed for John Deere equipment