Learn More
MSC Baldwin Fuel/Water Separator Element, 4 11/32 in OD, 21/32 in ID, 6 7/16 in Length
Catalog No. 5000508008
Encompass
This fuel/water separator element is designed to keep fuel clean and engines running at maximum efficiency by removing particulate and water contamination, which is crucial for modern fuel injection systems.
  • Keeps fuel clean and engines running at maximum efficiency.
  • Protects fuel systems that operate at higher pressures (up to 60,000 psi) and tighter tolerances.
  • Prevents abrasive wear caused by microscopic particles (as small as 2-3 microns).
Catalog No. 50-005-08008 Supplier MSC Supplier No. 55866149
This fuel/water separator element is designed to keep fuel clean and engines running at maximum efficiency by removing particulate and water contamination, which is crucial for modern fuel injection systems.
  • Keeps fuel clean and engines running at maximum efficiency.
  • Protects fuel systems that operate at higher pressures (up to 60,000 psi) and tighter tolerances.
  • Prevents abrasive wear caused by microscopic particles (as small as 2-3 microns).