Learn More
MSC Baldwin Fuel/Water Separator, Spin-on, 1-14 Thread, 4 Micron Nominal
Catalog No. 5000508122
Encompass
Modern fuel injection systems require fuel to be free of both particulate and water contamination. This fuel/water separator ensures fuel is clean and engines run at maximum efficiency.
  • Spin-on design with drain for easy installation and maintenance.
  • Removes particulate and water contamination from fuel.
  • Features an integral post seal.
  • Furnished with an ES24-D gasket and an integral G381-A gasket.
  • Offers a 4 micron nominal and 20 micron absolute rating.
  • Designed for applications in Case and International Equipment.
  • Thread size of 1-14.
  • Outside diameter measures 3 11/16 in (93.7 mm).
  • Length measures 6 3/16 in (157.2 mm).
Catalog No. 50-005-08122 Supplier MSC Supplier No. 55862494
Modern fuel injection systems require fuel to be free of both particulate and water contamination. This fuel/water separator ensures fuel is clean and engines run at maximum efficiency.
  • Spin-on design with drain for easy installation and maintenance.
  • Removes particulate and water contamination from fuel.
  • Features an integral post seal.
  • Furnished with an ES24-D gasket and an integral G381-A gasket.
  • Offers a 4 micron nominal and 20 micron absolute rating.
  • Designed for applications in Case and International Equipment.
  • Thread size of 1-14.
  • Outside diameter measures 3 11/16 in (93.7 mm).
  • Length measures 6 3/16 in (157.2 mm).