Learn More
MSC Baldwin Fuel/Water Separator Spin-on with Drain, M16 x 1.5
Catalog No. 5000508269
Encompass
This spin-on fuel filter functions as a fuel/water separator, designed for modern fuel injection systems. It ensures fuel is free of both particulate and water contamination, keeping engines running at maximum efficiency.
  • Fuel/water separator spin-on with drain design.
  • M16 x 1.5 thread size.
  • 3 1/32 in (77.0 mm) outside diameter.
  • 6 21/32 in (169.1 mm) length.
  • Attached gasket and furnished gasket included.
  • 5 absolute micron rating.
  • Suitable for Case equipment and Gehl loaders.
Catalog No. 50-005-08269 Supplier MSC Supplier No. 55863229
This spin-on fuel filter functions as a fuel/water separator, designed for modern fuel injection systems. It ensures fuel is free of both particulate and water contamination, keeping engines running at maximum efficiency.
  • Fuel/water separator spin-on with drain design.
  • M16 x 1.5 thread size.
  • 3 1/32 in (77.0 mm) outside diameter.
  • 6 21/32 in (169.1 mm) length.
  • Attached gasket and furnished gasket included.
  • 5 absolute micron rating.
  • Suitable for Case equipment and Gehl loaders.