Learn More
MSC Baldwin Fuel/Water Separator Spin-on, Drain and Sensor Port, 3/9 Micron
Catalog No. 5000508476
Encompass
This fuel/water separator spin-on is designed to ensure modern fuel injection systems receive fuel free from particulate and water contamination. It helps keep engines running at maximum efficiency by maintaining fuel cleanliness.
  • Designed to separate fuel from water and particulate contamination.
  • Includes a drain and sensor port.
  • Features an integral post seal.
  • Equipped with an integral gasket and included O-ring for a secure fit.
  • Provides 3 nominal and 9 absolute micron filtration.
Catalog No. 50-005-08476 Supplier MSC Supplier No. 55863831
This fuel/water separator spin-on is designed to ensure modern fuel injection systems receive fuel free from particulate and water contamination. It helps keep engines running at maximum efficiency by maintaining fuel cleanliness.
  • Designed to separate fuel from water and particulate contamination.
  • Includes a drain and sensor port.
  • Features an integral post seal.
  • Equipped with an integral gasket and included O-ring for a secure fit.
  • Provides 3 nominal and 9 absolute micron filtration.