Learn More
MSC Baldwin Fuel/Water Separator Spin-on, M14 x 1.5 Thread, 9 Micron
Catalog No. 5000509394
Encompass
This fuel/water separator spin-on filter is designed to remove particulate and water contamination from fuel, ensuring modern fuel injection systems operate effectively and engines run at maximum efficiency.
  • Removes particulate and water contamination from fuel
  • Features a spin-on design with an integral drain
  • M14 x 1.5 thread size for precise fitting
  • Outside diameter measures 3 11/16 in (93.7 mm)
  • Overall length of 7 11/32 in (186.5 mm)
  • Includes an integral gasket for a secure seal
  • Provides 9 micron nominal and 40 micron absolute filtration
  • Suitable for Iveco EuroCargo, EuroStar, and EuroTech series trucks
Catalog No. 50-005-09394 Supplier MSC Supplier No. 55862528
This fuel/water separator spin-on filter is designed to remove particulate and water contamination from fuel, ensuring modern fuel injection systems operate effectively and engines run at maximum efficiency.
  • Removes particulate and water contamination from fuel
  • Features a spin-on design with an integral drain
  • M14 x 1.5 thread size for precise fitting
  • Outside diameter measures 3 11/16 in (93.7 mm)
  • Overall length of 7 11/32 in (186.5 mm)
  • Includes an integral gasket for a secure seal
  • Provides 9 micron nominal and 40 micron absolute filtration
  • Suitable for Iveco EuroCargo, EuroStar, and EuroTech series trucks