Learn More
MSC Baldwin Maximum Performance Fuel/Water Separator Element, 5 Micron Absolute, 4 11/32 in OD, 6 1/2 in Height
Catalog No. 5000509655
Encompass
This is a maximum performance fuel/water separator element designed to efficiently separate fuel and water, ensuring clean fuel delivery. It features a 5 micron absolute rating for superior filtration and includes an O-ring and an attached grommet for easy installation and a secure fit.
  • Maximum performance fuel/water separation
  • 5 micron absolute filtration rating
  • Outside diameter: 4 11/32 in (110.3 mm)
  • Inside diameter: 21/32 in (16.7 mm)
  • Height: 6 1/2 in (165.1 mm)
  • Includes an O-ring
  • Features an attached grommet
Catalog No. 50-005-09655 Supplier MSC Supplier No. 55868798
This is a maximum performance fuel/water separator element designed to efficiently separate fuel and water, ensuring clean fuel delivery. It features a 5 micron absolute rating for superior filtration and includes an O-ring and an attached grommet for easy installation and a secure fit.
  • Maximum performance fuel/water separation
  • 5 micron absolute filtration rating
  • Outside diameter: 4 11/32 in (110.3 mm)
  • Inside diameter: 21/32 in (16.7 mm)
  • Height: 6 1/2 in (165.1 mm)
  • Includes an O-ring
  • Features an attached grommet