Learn More
MSC Baldwin Outer Air Element, 12 3/32 in OD, 22 1/2 in Length
Catalog No. 5000509691
Encompass
Baldwin axial seal air filter elements ensure proper equipment function by preventing airborne contaminants from reaching the combustion chamber.
  • Outer air element design
  • Outside diameter of 12 3/32 in (307.2 mm)
  • Inside diameter of 21/32 in (16.7 mm) and 7 11/16 in (195.3 mm)
  • Length of 22 1/2 in (571.5 mm)
  • Attached gasket for secure sealing
  • Suitable for John Deere equipment applications
Catalog No. 50-005-09691 Supplier MSC Supplier No. 55869267
Baldwin axial seal air filter elements ensure proper equipment function by preventing airborne contaminants from reaching the combustion chamber.
  • Outer air element design
  • Outside diameter of 12 3/32 in (307.2 mm)
  • Inside diameter of 21/32 in (16.7 mm) and 7 11/16 in (195.3 mm)
  • Length of 22 1/2 in (571.5 mm)
  • Attached gasket for secure sealing
  • Suitable for John Deere equipment applications